CH6371380140 Chem Impex International, Inc.

Fmoc-L-allo-threoninol

  • MDL number MFCD01318737
  • Cas Number : 252049-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-146
Synonym Fmoc-allo-L-Thr-ol
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-i-allo-i-threoninol/4914?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-i-allo-i-threoninol/4914?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart