CH6371380141 Chem Impex International, Inc.

Fmoc-N-methyl-L-threonine

  • MDL number MFCD02094575
  • Cas Number : 252049-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-159 �C
Synonym Fmoc-N-Me-L-Thr-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-l-threonine/4915?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-l-threonine/4915?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart