CH6371380145 Chem Impex International, Inc.

Fmoc-O-phospho-L-serine

  • MDL number MFCD00672324
  • Cas Number : 158171-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Ser(H2PO3)-OH
Purity 99
Storage temp. Store at = -4
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-phospho-l-serine/4920?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-phospho-l-serine/4920?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart