CH6371380147 Chem Impex International, Inc.

Fmoc-O-sulfo-L-tyrosine sodium salt

  • MDL number MFCD00077078
  • Cas Number : 106864-37-3
  • Molecular Weight : 505.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-sulfo-l-tyrosine-sodium-salt/4923?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-sulfo-l-tyrosine-sodium-salt/4923?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart