CH6371380148 Chem Impex International, Inc.

Formyl-L-alanine methyl ester

  • MDL number MFCD00080914
  • Cas Number : 32221-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym For-L-Ala-OMe
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/formyl-l-alanine-methyl-ester/4924?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/formyl-l-alanine-methyl-ester/4924?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart