CH6371380151 Chem Impex International, Inc.

For-L-norleucine

  • MDL number MFCD00237678
  • Cas Number : 133388-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-128
Synonym For-L-Nle-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/for-l-norleucine/4928?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/for-l-norleucine/4928?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart