CH6371380152 Chem Impex International, Inc.

Fromyl-L-Valine

  • MDL number MFCD00021029
  • Cas Number : 4289-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-158
Synonym For-L-Val-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fromyl-l-valine/4930?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fromyl-l-valine/4930?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart