CH6371380154 Chem Impex International, Inc.

Glutathione-monoisopropyl ester (reduced)

  • MDL number MFCD00237073
  • Cas Number : 97451-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym H-Glu(Cys-Gly-isopropyl ester)-OH
Purity 94
Storage temp. Store at =-4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glutathione-monoisopropyl-ester-reduced/4935?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glutathione-monoisopropyl-ester-reduced/4935?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart