CH6371380156 Chem Impex International, Inc.

g-Aminobutyric acid tert-butyl ester hydrochloride

  • MDL number MFCD00270592
  • Cas Number : 58640-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym gamma-Abu-OtBu HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gamma-aminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/4938?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gamma-aminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/4938?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart