CH6371380159 Chem Impex International, Inc.

Ala-Gly-NH2�

  • MDL number MFCD00190742
  • Cas Number : 71431-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Alanyl glycine amide hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-gly-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hcl/4943?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-gly-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hcl/4943?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart