CH6371380160 Chem Impex International, Inc.

Ala-Pro-NH2�

  • MDL number MFCD00237756
  • Cas Number : 76399-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4
Physical Form White to off-white solid or powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-pro-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hcl/4944?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-pro-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hcl/4944?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart