CH6371380163 Chem Impex International, Inc.

L-Aspartic acid b-benzyl ester 7-amido-4-methylcoumarin hydrochloride

  • MDL number MFCD01318835
  • Cas Number : 219138-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-201
Synonym L-Asp(OBzl)-AMC HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-beta-benzyl-ester-7-amido-4-methylcoumarin-hydrochloride/4949?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-beta-benzyl-ester-7-amido-4-methylcoumarin-hydrochloride/4949?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart