CH6371380165 Chem Impex International, Inc.

D-Leucine amide

  • MDL number MFCD00237134
  • Cas Number : 15893-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Leu-NH2
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-leucine-amide/4952?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-leucine-amide/4952?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart