CH6371380167 Chem Impex International, Inc.

DL-Leucine methyl ester hydrochloride

  • MDL number MFCD00067541
  • Cas Number : 6322-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-leucine-methyl-ester-hydrochloride/4955?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-leucine-methyl-ester-hydrochloride/4955?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart