CH6371380172 Chem Impex International, Inc.

DL-Phenylalanine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00035105
  • Cas Number : 119290-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Phe-OBzl TosOH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-phenylalanine-benzyl-ester-4-toluenesulfonate-salt/4961?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-phenylalanine-benzyl-ester-4-toluenesulfonate-salt/4961?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart