CH6371380177 Chem Impex International, Inc.

DL-tryptophan amide hydrochloride

  • MDL number MFCD00070409
  • Cas Number : 67607-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Trp-NH2 HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-tryptophan-amide-hydrochloride/4966?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-tryptophan-amide-hydrochloride/4966?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart