CH6371380178 Chem Impex International, Inc.

DL-Tryptophan ethyl ester hydrochloride

  • MDL number MFCD00067553
  • Cas Number : 6519-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Trp-OEt HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-tryptophan-ethyl-ester-hydrochloride/4967?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-tryptophan-ethyl-ester-hydrochloride/4967?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart