CH6371380179 Chem Impex International, Inc.

DL-Tryptophan methyl ester hydrochloride

  • MDL number MFCD00066133
  • Cas Number : 5619-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-219
Synonym DL-Trp-OMe�
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-tryptophan-methyl-ester-hydrochloride/4968?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-tryptophan-methyl-ester-hydrochloride/4968?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart