CH6371380180 Chem Impex International, Inc.

Nepsilon-Allyloxycarbonyl-D-lysine

  • MDL number MFCD02259523
  • Cas Number : 274260-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 255 °C (dec.)
Purity 98
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-allyloxycarbonyl-d-lysine/4969?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-allyloxycarbonyl-d-lysine/4969?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart