CH6371380181 Chem Impex International, Inc.

D-Norvaline benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD01318877
  • Cas Number : 218962-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141 - 147
Synonym D-Nva-OBzl TosOH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-norvaline-benzyl-ester-4-toluenesulfonate-salt/4971?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-norvaline-benzyl-ester-4-toluenesulfonate-salt/4971?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart