CH6371380182 Chem Impex International, Inc.

D-Val-D-Val-OH

  • MDL number MFCD00237311
  • Cas Number : 62653-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268 - 272 �dec.)
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-val-d-val-oh/4974?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-val-d-val-oh/4974?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart