CH6371380183 Chem Impex International, Inc.

Nd-4,4'-Dimethoxydityl-L-glutamine methyl ester hydrochloride

  • MDL number MFCD08272681
  • Cas Number : 28252-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-180
Synonym L-Gln(Dod)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-4-4-dimethoxydityl-l-glutamine-methyl-ester-hydrochloride/4976?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-4-4-dimethoxydityl-l-glutamine-methyl-ester-hydrochloride/4976?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart