CH6371380187 Chem Impex International, Inc.

Gly-Ala-Phe-OH

  • MDL number MFCD00038178
  • Cas Number : 17922-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycyl-L-alanyl-L-phenylalanine
Purity 99
Storage temp. Store at =-4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-ala-phe-oh/4983?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-ala-phe-oh/4983?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart