CH6371380189 Chem Impex International, Inc.

Gly-Gly-Ala-OH

  • MDL number MFCD00065113
  • Cas Number : 19729-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycyl-glycyl-L-alanine
Purity 0.95
Storage temp. Store at = -15
Physical Form Solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-gly-ala-oh/4989?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-gly-ala-oh/4989?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart