CH6371380192 Chem Impex International, Inc.

Gly-Gly-Gly-NH2�

  • MDL number MFCD00237928
  • Cas Number : 35790-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at =-4
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-gly-gly-nh-sub-2-sub-·hcl/4992?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-gly-gly-nh-sub-2-sub-·hcl/4992?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart