CH6371380194 Chem Impex International, Inc.

Gly-Gly-His-OH

  • MDL number MFCD00079323
  • Cas Number : 93404-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycylglycyl-L-histidine
Purity 96
Storage temp. Store at = - 4
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-gly-his-oh/4994?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-gly-his-oh/4994?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart