CH6371380198 Chem Impex International, Inc.

Gly-Gly-Phe-OH

  • MDL number MFCD00066054
  • Cas Number : 6234-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycyl-glycyl-L-phenylalanine
Purity 95
Storage temp. Store at =-4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-gly-phe-oh/4998?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-gly-phe-oh/4998?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart