CH6371380201 Chem Impex International, Inc.

Gly-Leu-NH2 HCl

  • MDL number MFCD00058254
  • Cas Number : 38173-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-213 °C
Purity 99
Storage temp. Store at =-4°C
Physical Form White product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-leu-nh-sub-2-sub-·hcl/5004?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-leu-nh-sub-2-sub-·hcl/5004?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart