CH6371380202 Chem Impex International, Inc.

Gly-Phe-Gly-OH

  • MDL number MFCD00083706
  • Cas Number : 14656-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycyl-L-phenylalanyl-glycine
Purity 98
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-phe-gly-oh/5005?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-phe-gly-oh/5005?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart