CH6371380203 Chem Impex International, Inc.

Gly-Phe-pNA

  • MDL number MFCD00038182
  • Cas Number : 21027-72-5
  • Molecular Weight : 342.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-187 �C
Purity 99
Storage temp. Store at 0-8
Physical Form Slightly yellowish to white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/gly-phe-pna/5006?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-phe-pna/5006?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart