CH6371380205 Chem Impex International, Inc.

Glycine b-naphthylamide

  • MDL number MFCD00046461
  • Cas Number : 716-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 124
Synonym Gly-betaNA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glycine-ß-naphthylamide/5009?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glycine-ß-naphthylamide/5009?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart