CH6371380206 Chem Impex International, Inc.

Gly-Thr-OH

  • MDL number MFCD00038176
  • Cas Number : 7093-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2S,3R)-2-(2-Aminoacetamido)-3-hydroxybutanoic acid
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-thr-oh/5010?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-thr-oh/5010?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart