CH6371380209 Chem Impex International, Inc.

His-Leu-OH

  • MDL number MFCD00037860
  • Cas Number : 7763-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Histidyl-L-leucine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/his-leu-oh/5013?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/his-leu-oh/5013?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart