CH6371380210 Chem Impex International, Inc.

Ile-Gly-Gly-OH

  • MDL number MFCD00037784
  • Cas Number : 68293-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Isoleucylglycylglycine
Purity 98
Storage temp. Store at =-4
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ile-gly-gly-oh/5014?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ile-gly-gly-oh/5014?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart