CH6371380211 Chem Impex International, Inc.

Leu-Gly-Leu-OH

  • MDL number MFCD00038310
  • Cas Number : 19408-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at =-4°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/leu-gly-leu-oh/5015?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/leu-gly-leu-oh/5015?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart