CH6371380212 Chem Impex International, Inc.

Leu-Gly-NH2�

  • MDL number MFCD00190943
  • Cas Number : 28671-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Leucylglycine amide hydrobromide
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/leu-gly-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hbr/5016?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/leu-gly-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hbr/5016?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart