CH6371380214 Chem Impex International, Inc.

Leu-Leu-Val-Phe-OH

  • MDL number MFCD00238025
  • Cas Number : 24205-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/leu-leu-val-phe-oh/5018?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/leu-leu-val-phe-oh/5018?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart