CH6371380218 Chem Impex International, Inc.

Lys-Ala-bNA

  • MDL number MFCD00038702
  • Cas Number : 20314-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-114
Synonym Lys-Ala-beta-naphthylamide
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/lys-ala-ßna/5027?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/lys-ala-ßna/5027?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart