CH6371380222 Chem Impex International, Inc.

L-Lysine b-naphtylamide carbonate

  • MDL number MFCD00051300
  • Cas Number : 18905-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-155
Synonym L-Lys-betaNA carbonate salt
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-lysine-beta-naphtylamide-carbonate/5035?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-lysine-beta-naphtylamide-carbonate/5035?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart