CH6371380226 Chem Impex International, Inc.

L-Phenylalanine 7-amido-4-methylcoumarin trifluoroacetate

  • MDL number MFCD00167024
  • Cas Number : 108321-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe-AMC TFA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-phenylalanine-7-amido-4-methylcoumarin-trifluoroacetate/5039?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-phenylalanine-7-amido-4-methylcoumarin-trifluoroacetate/5039?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart