CH6371380227 Chem Impex International, Inc.

Phe-Arg-bNA�l

  • MDL number MFCD00058046
  • Cas Number : 100929-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Phe-Arg beta-naphthylamide dihydrochloride
Purity 99
Storage temp. Store at =-4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phe-arg-ßna·2hcl/5040?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phe-arg-ßna·2hcl/5040?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart