CH6371380228 Chem Impex International, Inc.

Z-L-valine trifluoroethyl ester

  • MDL number MFCD03788136
  • Cas Number : 203640-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-61
Synonym Z-L-Val-OTfa
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-valine-trifluoroethyl-ester/5041?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-valine-trifluoroethyl-ester/5041?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart