CH6371380229 Chem Impex International, Inc.

Phe-Leu-NH2�

  • MDL number MFCD00058006

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phe-leu-nh-sub-2-sub-b-·-b-hcl/5042?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phe-leu-nh-sub-2-sub-b-·-b-hcl/5042?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart