CH6371380230 Chem Impex International, Inc.

H-D-His(3-Me)-OH

  • MDL number MFCD00067633
  • Cas Number : 163750-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-His(p-Me)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Slightly yellowish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/h-d-his-3-me-oh/5043?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-d-his-3-me-oh/5043?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart