CH6371380233 Chem Impex International, Inc.

Pro-Pro-OH�

  • MDL number MFCD00191008
  • Cas Number : 76932-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Prolyl-L-proline hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pro-pro-oh·hcl/5046?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pro-pro-oh·hcl/5046?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart