CH6371380234 Chem Impex International, Inc.

Pro-Trp-OH

  • MDL number MFCD00037943
  • Cas Number : 35310-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Prolyl-L-tryptophan
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pro-trp-oh/5047?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pro-trp-oh/5047?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart