CH6371380235 Chem Impex International, Inc.

Pro-Tyr-OH

  • MDL number MFCD00037870
  • Cas Number : 19786-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-210
Synonym L-Prolyl-L-tyrosine
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pro-tyr-oh/5048?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pro-tyr-oh/5048?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart