CH6371380236 Chem Impex International, Inc.

O-Acetyl-L-serine hydrochloride

  • MDL number MFCD00060169
  • Cas Number : 66638-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ser(Ac)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-acetyl-l-serine-hydrochloride/5050?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-acetyl-l-serine-hydrochloride/5050?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart