CH6371380238 Chem Impex International, Inc.

Ser-Gly-Gly-OH

  • MDL number MFCD00238167
  • Cas Number : 2543-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ser-Gly-Gly-OH
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ser-gly-gly-oh/5053?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ser-gly-gly-oh/5053?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart