CH6371380239 Chem Impex International, Inc.

Ser-Gly-OH

  • MDL number MFCD00037778
  • Cas Number : 687-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Seryl-L-glycine
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ser-gly-oh/5054?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ser-gly-oh/5054?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart